Learn More
Analytical Sales & Services 9mm, 2mL Clear Wide Mouth Twist Vials and Blue Caps with Pre-Slit PTFE/Silicone Liners
Catalog No. 5000805591
Encompass
Advantage™ Unassembled Vial Kit. Contains: 9mm, 2mL Clear Glass Wide Mouth Twist Vials and 9mm Blue Twist Caps with Pre-Slit PTFE/Silicone Liners
Catalog No. 50-008-05591 Supplier Analytical Sales & Services Supplier No. 88535SLCASE
Advantage™ Unassembled Vial Kit. Contains: 9mm, 2mL Clear Glass Wide Mouth Twist Vials and 9mm Blue Twist Caps with Pre-Slit PTFE/Silicone Liners