Learn More
Analytical Sales & Services 9mm, 2mL Clear Wide Mouth Twist Vials and Blue Caps with PTFE/Silicone Liners
Catalog No. 5000804595
Encompass
Advantage™ Pre-Assembled Vial Kit. Contains: 9mm, 2mL Clear Glass Wide Mouth Twist Vials with Step and 9mm Blue Twist Caps with PTFE/Silicone Liners
Catalog No. 50-008-04595 Supplier Analytical Sales & Services Supplier No. 58128CASE
Advantage™ Pre-Assembled Vial Kit. Contains: 9mm, 2mL Clear Glass Wide Mouth Twist Vials with Step and 9mm Blue Twist Caps with PTFE/Silicone Liners