Learn More
Analytical Sales & Services 9mm, 2mL Clear Wide Mouth Twist Vials and Blue Caps with PTFE/Silicone/PTFE Liners
Catalog No. 5000805556
Encompass
Advantage™ Unassembled Vial Kit. Contains: 9mm, 2mL Clear Glass Wide Mouth Twist Vials and 9mm Blue Twist Caps with PTFE/Silicone/PTFE Liners
Catalog No. 50-008-05556 Supplier Analytical Sales & Services Supplier No. 88536CASE
Advantage™ Unassembled Vial Kit. Contains: 9mm, 2mL Clear Glass Wide Mouth Twist Vials and 9mm Blue Twist Caps with PTFE/Silicone/PTFE Liners