Learn More
Analytical Sales & Services 9mm, 2mL Clear Wide Mouth Twist Vials with Marking Spot and Blue Caps with Pre-Slit Silicone/PTFE Liners
Catalog No. 5000805064
Encompass
Advantage™ Unassembled Vial Kit. Contains: 9mm, 2mL Clear Glass Wide Mouth Twist Vials with Marking Spot and 9mm Blue Twist Caps with Pre-Slit Silicone/PTFE Liners
Catalog No. 50-008-05064 Supplier Analytical Sales & Services Supplier No. 58088CASE
Advantage™ Unassembled Vial Kit. Contains: 9mm, 2mL Clear Glass Wide Mouth Twist Vials with Marking Spot and 9mm Blue Twist Caps with Pre-Slit Silicone/PTFE Liners