Learn More
Analytical Sales & Services 9mm, 2mL Clear Wide Mouth Twist Vials with Marking Spot and Blue Caps with Silicone/PTFE Liners
Catalog No. 5000804654
Encompass
Advantage™ Unassembled Vial Kit. Contains: 9mm, 2mL Clear Glass Wide Mouth Twist Vials with Marking Spot and 9mm Blue Twist Caps with Silicone/PTFE Liners
Catalog No. 50-008-04654 Supplier Analytical Sales & Services Supplier No. 58077CASE
Advantage™ Unassembled Vial Kit. Contains: 9mm, 2mL Clear Glass Wide Mouth Twist Vials with Marking Spot and 9mm Blue Twist Caps with Silicone/PTFE Liners