Learn More
Analytical Sales & Services 9mm, 2mL Clear Wide Mouth Twist Vials with Marking Spot and Blue Tight Fit Caps with Silicone/PTFE Liners. *Direct Agilent Replacement.
Catalog No. 5000806516
Encompass
Advantage™ Unassembled Vial Kit. Contains: 9mm, 2mL Clear Glass Wide Mouth Twist Vials with Marking Spot and 9mm Blue Tight Fit Caps with Silicone/PTFE Liners. *Direct Agilent Replacement.
Catalog No. 50-008-06516 Supplier Analytical Sales & Services Supplier No. 868127CASE
Advantage™ Unassembled Vial Kit. Contains: 9mm, 2mL Clear Glass Wide Mouth Twist Vials with Marking Spot and 9mm Blue Tight Fit Caps with Silicone/PTFE Liners. *Direct Agilent Replacement.