Learn More
SAI Infusion Technologies AutoClip Wound Clips - 9mm, 1000/box
SDP
Catalog No. NC2160653
Encompass
Use these staple-like 9mm AutoClip Wound Clips to quickly and cleanly close wounds following surgery. Clips are applied with the 9mm AutoClip Applier and can be removed with the 9mm AutoClip Remover (Applier and Remover are sold separately). Stainless steel and non-pyrogenic. 1000/box, Non-Sterile. Can be autoclaved. Product is for laboratory animal research only.
This item is not returnable. View return policy
Catalog No. NC2160653 Supplier SAI Infusion Technologies Supplier No. ACWC9MM
Use these staple-like 9mm AutoClip Wound Clips to quickly and cleanly close wounds following surgery. Clips are applied with the 9mm AutoClip Applier and can be removed with the 9mm AutoClip Remover (Applier and Remover are sold separately). Stainless steel and non-pyrogenic. 1000/box, Non-Sterile. Can be autoclaved. Product is for laboratory animal research only.