Learn More
GENTECH SOLUTIONS LLC 9mm Blue Open Top Screw Thread Cap, Bonded 0.040" Red Silicone Rubber/Clear PTFE
Catalog No. NC3728810
Encompass_Preferred
EliteChem Made in the USA with the highest purity silicone and premium-quality PTFE films to produce the Septum in the closures. Compatible with chromatography vials with a 9-425 closure size.
Catalog No. NC3728810 Supplier GENTECH SOLUTIONS LLC Supplier No. 2000200010
EliteChem Made in the USA with the highest purity silicone and premium-quality PTFE films to produce the Septum in the closures. Compatible with chromatography vials with a 9-425 closure size.