Learn More
Chemglass Life Sciences Preassembled Ribbed Closure and Septa, Yellow, 9mm Thread, PTFE/Red Rubber Septa
SDP
Catalog No. 501215217
Encompass_Preferred
Preassembled Ribbed Closure and Septa, Yellow, 9mm Thread, PTFE/Red Rubber Septa. Pre-assembled closure and septa minimize contamination from handling. They are convenient and available in a variety of colors with your choice of septa.
Catalog No. 50-121-5217 Supplier Chemglass Life Sciences Supplier No. CV3791Y009
Preassembled Ribbed Closure and Septa, Yellow, 9mm Thread, PTFE/Red Rubber Septa. Pre-assembled closure and septa minimize contamination from handling. They are convenient and available in a variety of colors with your choice of septa.