Learn More
Braintree Scientific 9mm EZ Clip Applier
SDP
Supplier:  Braintree Scientific EZCAPL
Encompass
The EZ Clip™ Ideal for use with lab animals or in veterinary surgery. Racks of 9mm clips are individually packaged in heat sealed, peel-away sleeves for cleanliness, ease of handling and storage. EZ Clips™ are packaged 20 clips on a rack, with 5 racks per strip (100 clips total). They can be purchased by the strip, 2 strips (200 clips) or by the case (1000 clips). Clips are pyrogen free and non-sterile. However, the autoclavable plastic packaging allows for sterilization of the clips without removing them from the packaging. The EZ Clip Applier is a stainless steel applying forcep that holds one rack of 20 clips. Works like a staple gun to apply clips to wound. The clips are fed by rustless beryllium-copper springs. The EZ Clip Remover™ is also stainless steel and can be used to remove all sizes of wound clips. **Sold for research purposes only and not for human use**
Catalog No. 50-195-4805