Learn More
Chemglass Life Sciences #9 Quartz O-Ring Joint, 12.0mm Tube O.D., Clamp Size/18, 112 O-Ring Size
SDP
Catalog No. CGQ013503
Encompass_Preferred
#9 Quartz O-Ring Joint, 12.0mm Tube O.D., Clamp Size/18, 112 O-Ring Size. Clear fused quartz o-ring joint. Mating surface is formed so that quartz joints can be connected directly to CG-124 borosilicate o-ring joints. For clamps see CG-150.
Catalog No. CGQ013503 Supplier Chemglass Life Sciences Supplier No. CGQ013503
#9 Quartz O-Ring Joint, 12.0mm Tube O.D., Clamp Size/18, 112 O-Ring Size. Clear fused quartz o-ring joint. Mating surface is formed so that quartz joints can be connected directly to CG-124 borosilicate o-ring joints. For clamps see CG-150.