Learn More
Ace Glass, Inc. 9MM ID GRADED SEALS 9MM ID GRADED SEALS
SDP
Catalog No. 50974594
Encompass
9mm quartz graded seals for connecting Pyrex or Kimax borosilicate glass to Quartz glass. Quartz length is 7.6cm, and borosilicate length is 10.2cm.
Catalog No. 50-974-594 Supplier Ace Glass, Inc. Supplier No. 845514
9mm quartz graded seals for connecting Pyrex or Kimax borosilicate glass to Quartz glass. Quartz length is 7.6cm, and borosilicate length is 10.2cm.