Learn More
Ace Glass, Inc. 9MM JOINTS O RING SEAL 112 9MM JOINTS O RING SEAL 112
SDP
Catalog No. NC0843644
Encompass_Preferred
9mm glass o-ring seal joint tooled with a groove to take a -112 FETFE o-ring, and to provide a greaseless, vacuum-tight connection. Supplied with one o-ring, which may be used up to 230¿C.
Catalog No. NC0843644 Supplier Ace Glass, Inc. Supplier No. 764606
9mm glass o-ring seal joint tooled with a groove to take a -112 FETFE o-ring, and to provide a greaseless, vacuum-tight connection. Supplied with one o-ring, which may be used up to 230¿C.