Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vetequip Inc 9MM NOSE CONE
SDP

Catalog No. NC9951444
Encompass_Preferred

Circuit, Rodent, NRB, w/ 09mm Nose Cone

Catalog No. NC9951444 Supplier Vetequip Inc Supplier No. 921461
May include imposed supplier surcharges.
 Restricted Item
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Circuit, Rodent, NRB, w/ 09mm Nose Cone

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.