Learn More
JG FINNERAN 9MM R.A.M. SMTH GRN CAP,1000/C
Catalog No. NC1964383
Encompass_Preferred
9mm R.A.M.™ Smooth Green Cap, PTFE/Butyl Rubber
Catalog No. NC1964383 Supplier JG FINNERAN Supplier No. 5394F09G
9mm R.A.M.™ Smooth Green Cap, PTFE/Butyl Rubber