Learn More
Cayman Chemical 9Nitrooleate Fatty Acids and D
SDP
Catalog No. 502888607
Encompass_Preferred
A novel endogenous lipid signaling molecule; nitration product of oleic acid in vivo mediated by peroxynitrite, acidified nitrite, or myeloperoxidase (in the presence of H2O2 and nitrite)
Catalog No. 50-288-8607 Supplier Cayman Chemical Supplier No. 10008042500 UG
A novel endogenous lipid signaling molecule; nitration product of oleic acid in vivo mediated by peroxynitrite, acidified nitrite, or myeloperoxidase (in the presence of H2O2 and nitrite)