Learn More
Cayman Chemical 9Nitrooleate Fatty Acids and D
SDP
Catalog No. 502887702
Encompass_Preferred
A novel endogenous lipid signaling molecule; nitration product of oleic acid in vivo mediated by peroxynitrite, acidified nitrite, or myeloperoxidase (in the presence of H2O2 and nitrite)
Catalog No. 50-288-7702 Supplier Cayman Chemical Supplier No. 10008042100 UG
A novel endogenous lipid signaling molecule; nitration product of oleic acid in vivo mediated by peroxynitrite, acidified nitrite, or myeloperoxidase (in the presence of H2O2 and nitrite)