Learn More
Cayman Chemical 9Nitrooleated17 GC MS Standard
SDP
Supplier:  Cayman Chemical 10557500 UG
Encompass_Preferred
An internal standard for the quantification of resolvin D1 by GC- or LC-MS
Catalog No. 50-288-7328