Learn More
Cayman Chemical 9Nitrooleated17 GC MS Standard
SDP
Catalog No. 502887627
Encompass_Preferred
An internal standard for the quantification of resolvin D1 by GC- or LC-MS
Catalog No. 50-288-7627 Supplier Cayman Chemical Supplier No. 105571 MG
An internal standard for the quantification of resolvin D1 by GC- or LC-MS