Learn More
Cayman Chemical 9OxoOTrE Fatty Acids and Deriv
SDP
Catalog No. 502888856
Encompass_Preferred
9-OxoOTrE is produced by the oxidation of 9-HpOTrE.{14394} 9-OxoOTrE exhibits antimicrobial activity against plant pathogenic microorganisms including bacteria and fungi.{14395}
Catalog No. 50-288-8856 Supplier Cayman Chemical Supplier No. 10009215250 UG
9-OxoOTrE is produced by the oxidation of 9-HpOTrE.{14394} 9-OxoOTrE exhibits antimicrobial activity against plant pathogenic microorganisms including bacteria and fungi.{14395}