Learn More
Cayman Chemical 9OxoOTrE Fatty Acids and Deriv
SDP
Supplier:  Cayman Chemical 1000921525 UG
Encompass_Preferred
9-OxoOTrE is produced by the oxidation of 9-HpOTrE.{14394} 9-OxoOTrE exhibits antimicrobial activity against plant pathogenic microorganisms including bacteria and fungi.{14395}
Catalog No. 50-288-8118