Learn More
USA Scientific Inc 9-place upright freezer rk
Catalog No. 502745005
Encompass
9-place (3 long x 3 high) upright freezer rack for 2 in H boxes, 16 5/16 in L x 5 1/2 in W x 6 5/8 in H, stainless steel
Catalog No. 50-274-5005 Supplier USA Scientific Inc Supplier No. 26023300
9-place (3 long x 3 high) upright freezer rack for 2 in H boxes, 16 5/16 in L x 5 1/2 in W x 6 5/8 in H, stainless steel