Learn More
Cole-Parmer Cryopak Ice-Pak Reusable Gel-Filled Ice Packs; 0 degrees C, 9" x 6-1/2" x 1" cs/12
Catalog No. nc1280846 Shop All Cole-Parmer Products
Encompass_Preferred
The refrigerant is designed to protect your product from freezing or over-heating even in extreme conditions. Gel filling is nontoxic, nonhazardous and biodegradable; won't contaminate samples. Reusable packs freeze solid in 6 to 10 hours and remain at constant temperature for up to 72 hours. Place packs in the freezer for 24 hours before use.
Catalog No. NC1280846 Supplier Cole-Parmer Supplier No. 0634605
The refrigerant is designed to protect your product from freezing or over-heating even in extreme conditions. Gel filling is nontoxic, nonhazardous and biodegradable; won't contaminate samples. Reusable packs freeze solid in 6 to 10 hours and remain at constant temperature for up to 72 hours. Place packs in the freezer for 24 hours before use.