Learn More
Andwin Scientific 9X9 SPECWIPE 300/PK
SDP
Catalog No. NC9512138
Encompass_Preferred
The Spec-Wipe 3 are ideal for sanitizing critical environments, for absorbing spills of water and most common solvents, and for other applications such as cleaning laminar flow benches. This product is made with 45% polyester and 55% cellulose and is measured to have a width of 22.9 centimeters and length of 22.9 centimeters. It is sold in a pack of 300.
This item is not returnable. View return policy
Catalog No. NC9512138 Supplier Andwin Scientific Supplier No. 21914758
The Spec-Wipe 3 are ideal for sanitizing critical environments, for absorbing spills of water and most common solvents, and for other applications such as cleaning laminar flow benches. This product is made with 45% polyester and 55% cellulose and is measured to have a width of 22.9 centimeters and length of 22.9 centimeters. It is sold in a pack of 300.