Learn More
Medchemexpress LLC HY-101735 50mg Medchemexpress, (9Z,11E)-Tetradecadien-1-yl acetate CAS:50767-79-8 Purity:>98%
SDP
Catalog No. 501932203
Encompass_Preferred
Medchemexpress, HY-101735 50mg (9Z,11E)-Tetradecadien-1-yl acetate CAS:50767-79-8 (9Z,11E)-Tetradecadien-1-yl acetate is the main component of the sex pheromone of female Spodoptera littoralis. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2203 Supplier Medchemexpress LLC Supplier No. HY10173550MG
Medchemexpress, HY-101735 50mg (9Z,11E)-Tetradecadien-1-yl acetate CAS:50767-79-8 (9Z,11E)-Tetradecadien-1-yl acetate is the main component of the sex pheromone of female Spodoptera littoralis. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.