Learn More
Medchemexpress LLC (9Z,11E)-tetradecadien-1-yl acetate | 50767-79-8 | MFCD00009871 | >98.0% | 252.39 | C16H28O2 | 100 MG
SDP
Catalog No. 5000246845
Encompass_Preferred
(9Z,11E)-Prodlure ((9Z,11E)-tetradecadien-1-yl acetate, CAS 50767-79-8) is an analytical reference standard of the primary sex pheromone component produced by female Spodoptera littoralis. Supplied as a neat liquid, it is intended for research and analytical applications, and is commonly used as a reference in chromatographic and mass spectrometric methods for identification and quantification.
  • Analytical reference standard for pheromone research.
  • Suitable for qualitative and quantitative HPLC, GC, and MS analyses.
  • Molecular formula C16H28O2; molecular weight 252.39.
  • Provided in small lab-scale pack sizes for analytical use.
  • Used in chemical ecology and insect behavior studies as a diagnostic standard.
Catalog No. 50-002-46845 Supplier Medchemexpress LLC Supplier No. HY101735100MG
(9Z,11E)-Prodlure ((9Z,11E)-tetradecadien-1-yl acetate, CAS 50767-79-8) is an analytical reference standard of the primary sex pheromone component produced by female Spodoptera littoralis. Supplied as a neat liquid, it is intended for research and analytical applications, and is commonly used as a reference in chromatographic and mass spectrometric methods for identification and quantification.
  • Analytical reference standard for pheromone research.
  • Suitable for qualitative and quantitative HPLC, GC, and MS analyses.
  • Molecular formula C16H28O2; molecular weight 252.39.
  • Provided in small lab-scale pack sizes for analytical use.
  • Used in chemical ecology and insect behavior studies as a diagnostic standard.