Learn More
Medchemexpress LLC (9Z,11E)-tetradecadien-1-yl acetate | 50767-79-8 | MFCD00009871 | 98.8% | 252.39 g/mol | C16H28O2 | 1 ML
SDP
Catalog No. 5000245526
Encompass_Preferred
(9Z,11E)-Tetradecadien-1-yl acetate (Prodlure) is a cis-trans unsaturated acetate ester used as a chemical ecology reagent. It is the primary sex-pheromone component of female Spodoptera littoralis and is supplied for analytical and behavioral research applications, typically as a 10 mM solution in DMSO or as a purified solid.
  • Used in pheromone biology and behavioral assays.
  • High purity (~98.8%) suitable for analytical work.
  • Available as a 10 mM DMSO solution or solid pack sizes for flexibility.
  • Serves as an analytical reference standard for identification of pheromone components.
  • Compatible with laboratory assays and chromatographic analysis.
Catalog No. 50-002-45526 Supplier Medchemexpress LLC Supplier No. HY10173510MM 1ML
(9Z,11E)-Tetradecadien-1-yl acetate (Prodlure) is a cis-trans unsaturated acetate ester used as a chemical ecology reagent. It is the primary sex-pheromone component of female Spodoptera littoralis and is supplied for analytical and behavioral research applications, typically as a 10 mM solution in DMSO or as a purified solid.
  • Used in pheromone biology and behavioral assays.
  • High purity (~98.8%) suitable for analytical work.
  • Available as a 10 mM DMSO solution or solid pack sizes for flexibility.
  • Serves as an analytical reference standard for identification of pheromone components.
  • Compatible with laboratory assays and chromatographic analysis.