Learn More
Medchemexpress LLC (9Z)-9-octadecen-17-ynoic acid | 151333-45-8 | MFCD32697053 | 100.0% | 278.43 g/mol | C18H30O2 | 5 MG
SDP
Catalog No. 5000264107
Encompass_Preferred
Oleic acid alkyne is an oleic acid analog bearing a terminal alkyne functional group, supplied as a purified research reagent for bioorthogonal labeling, conjugation, and lipid metabolism studies. The terminal alkyne enables efficient copper-catalyzed azide-alkyne cycloaddition (click chemistry) to covalently attach probes or tags to lipid species. Handle and store according to supplier guidance.
  • Enables click chemistry for lipid labeling and tracking.
  • High purity for reliable experimental results.
  • Available in small milligram quantities for method development.
  • Soluble in common organic solvents such as DMSO.
  • For research use only; not for human or diagnostic use.
Catalog No. 50-002-64107 Supplier Medchemexpress LLC Supplier No. HY1271125MG
Oleic acid alkyne is an oleic acid analog bearing a terminal alkyne functional group, supplied as a purified research reagent for bioorthogonal labeling, conjugation, and lipid metabolism studies. The terminal alkyne enables efficient copper-catalyzed azide-alkyne cycloaddition (click chemistry) to covalently attach probes or tags to lipid species. Handle and store according to supplier guidance.
  • Enables click chemistry for lipid labeling and tracking.
  • High purity for reliable experimental results.
  • Available in small milligram quantities for method development.
  • Soluble in common organic solvents such as DMSO.
  • For research use only; not for human or diagnostic use.