Learn More
Cayman Chemical 9(Z),11(Z)-Conjugated Linoleic Acid methyl ester, Purity: ≥96%, CAS: 822-10-6, 25 mg
SDP
Catalog No. 502934420
Encompass_Preferred
A conjugated linoleic acid methyl ester; has been used as a standard for the quantification of conjugated linoleic acid methyl esters by GC-MS
Catalog No. 50-293-4420 Supplier Cayman Chemical Supplier No. 2458325 MG
A conjugated linoleic acid methyl ester; has been used as a standard for the quantification of conjugated linoleic acid methyl esters by GC-MS