Learn More
Cayman Chemical 9(Z),11(Z)-Conjugated Linoleic Acid, Purity: ≥96%, CAS: 544-70-7, 25 mg
SDP
Catalog No. 502934612
Encompass_Preferred
An isomer of linoleic acid; has been found in milk from cows and other animals; cytotoxic to Caco-2 human colon cancer cells (EC50 = 446.1 µM) and induces changes in the miRNA profile
Catalog No. 50-293-4612 Supplier Cayman Chemical Supplier No. 2458225 MG
An isomer of linoleic acid; has been found in milk from cows and other animals; cytotoxic to Caco-2 human colon cancer cells (EC50 = 446.1 µM) and induces changes in the miRNA profile