Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Revos™ Tissue Processor

Catalog No. a84100001a Shop All Epredia Products
Change view
Click to view available options
Voltage:
100/240 V
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Voltage
A84100001A 100/240 V
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A84100001A Supplier Epredia™ Supplier No. A84100001ATC
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The Epredia™ Revos™ workflow-enhancing tissue processor overcomes common workflow challenges. It offers reduced tissue processing time and less risk of tissue damage – all with no reduction in processing quality.

Rapid processing

Our unique, canted chamber enhances the distribution of reagent, reduces tissue processing time and allows for high-quality processing results.

Intelligent monitoring

The Revos' intelligent reagent management system tracks and logs reagent usage. It remembers every bottle loaded, protocol executed and service performed – ensuring your laboratory uses reagents to their fullest potential.

Sample safety

RFID technology enables Revos to sense where the 100% processing alcohol is loaded in the system and prevents incorrect replenishment.

Safer environment

The Revos' dual-filtration system and down-draft ventilation helps protect laboratory staff by controlling both solvent and formalin fumes.

Protecting tissue

Revos is ready for unexpected power outages, the on board battery backup system keep your samples safe and protocols active for up to three hours.

Enhanced serviceability

Advanced remote diagnostic capabilities are available to help minimize downtime and keep your lab running.

A smarter solution

The accuracy of patient tests often hinges upon proper preparation and consistency in processes throughout the laboratory.

When paired with Epredia™ Revos reagents featuring Epredia™ Syntri Safeguard™ RFID technology, the Revos intelligent reagent management system tracks and logs reagent usage. It remembers every bottle loaded, protocol executed and service performed – ensuring your laboratory uses reagents to their fullest potential.

  • Backup battery: 4 hours
  • Number of retorts: 1
  • Cassette capacity: Standard 300, Mega 42
  • Data connection: LIMS connectivity via RS232 port

Specifications

Product Type Tissue Processor
Reagent Capacity Alcohol configuration: 6 bottles; Xylene/XyleneSubstitute configuration: 3 bottles; Fixation configuration: 2 bottles
Certifications/Compliance IEC61010 Third Ed., UL
Depth (Metric) 580 mm
Height (Metric) 1370 mm
Width (Metric) 850 mm
Weight (Metric) Instrument: 165 kg; Instrument + typical reagents: 240 kg
Amperage 0.6 A ±10%
Frequency 50/60 Hz
Voltage 100/240 V
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.