Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

o-Tolyl isothiocyanate, 98%

Catalog No. AAB2284514
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2284514 25 g
AAB2284506 5 g
AAB2284522 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2284514 Supplier Thermo Scientific Chemicals Supplier No. B2284514
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 614-69-7 | C8H7NS | 149.211 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 614-69-7
Molecular Formula C8H7NS
Molecular Weight (g/mol) 149.211
MDL Number MFCD00004802
InChI Key JYKYYPPZLPVIBY-UHFFFAOYSA-N
Synonym 2-methylphenyl isothiocyanate, o-tolyl isothiocyanate, 2-tolyl isothiocyanate, o-tolylisothiocyanate, benzene, 1-isothiocyanato-2-methyl, 1-isothiocyanato-2-methyl-benzene, 2-methylphenylisothiocyanate, isothiocyanic acid o-tolyl ester, isothiocyanic acid, o-tolyl ester, 2-methylbenzenisothiocyanate
PubChem CID 69195
IUPAC Name 1-isothiocyanato-2-methylbenzene
SMILES CC1=CC=CC=C1N=C=S

Specifications

Density 1.115
Boiling Point 239°C
Flash Point 107°C (224°F)
Odor Pungent
Refractive Index 1.6361
Quantity 25 g
UN Number UN2922
Beilstein 878487
Sensitivity Moisture sensitive
Formula Weight 149.22
Percent Purity 98%
Chemical Name or Material o-Tolyl isothiocyanate
Show More Show Less
missing translation for 'healthHazard1' H302+H312+H332-H317-H334
missing translation for 'healthHazard2' GHS H Statement
H331-H314-H318-H302-H312
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
missing translation for 'healthHazard3' P261-P264b-P270-P271-P272-P280-P285-P301+P312-P302+P352-P304+P340-P312-P330-P333+P313-P342+P311-P363-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: II
EINECSNumber 210-390-6
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.