Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diethylenetriamine, 98+%

Catalog No. AC114310010
Change view
Click to view available options
Quantity:
250 mL
1 L
2.5 L
10 L
Packaging:
Glass bottle
Plastic drum
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC114310010 1 L Glass bottle
AC114312500 250 mL Glass bottle
AC114310025 2.5 L Glass bottle
AC114310100 10 L Plastic drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AC114310010 Supplier Thermo Scientific Chemicals Supplier No. 114310010
Only null left
Add to Cart

Diethylenetriamine, 98+%, C4H13N3, CAS Number-111-40-0, 1,4,7-triazaheptane, 2,2'-diaminodiethylamine, diethylenetriamine, 2,2'-iminodiethylamine, bis 2-aminoethyl amine, barsamide 115, ancamine deta, n,n-bis 2-aminoethyl amine, epicure t, diethylene triamine, 2.5L, 98% min. (GC), 04,255 | CAS: 111-40-0 | C4H13N3 | 103.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 111-40-0
Molecular Formula C4H13N3
Molecular Weight (g/mol) 103.17
MDL Number MFCD00008171
InChI Key RPNUMPOLZDHAAY-UHFFFAOYSA-N
Synonym diethylenetriamine, bis 2-aminoethyl amine, 2,2'-diaminodiethylamine, diethylene triamine, barsamide 115, epicure t, ancamine deta, 1,4,7-triazaheptane, 2,2'-iminodiethylamine, n,n-bis 2-aminoethyl amine
PubChem CID 8111
ChEBI CHEBI:30629
SMILES NCCNCCN

Specifications

Melting Point -40.0°C
Density 0.9600g/mL
Boiling Point 206.0°C
Flash Point 96°C
Infrared Spectrum Authentic
Assay Percent Range 98% min. (GC)
Packaging Glass bottle
Refractive Index 1.4826 to 1.4846
Linear Formula H2NCH2CH2NHCH2CH2NH2
Quantity 1 L
Beilstein 04,255
Specific Gravity 0.96
Solubility Information Solubility in water: miscible. Other solubilities: soluble in most common organic solvents, soluble in hydrocarbons
IUPAC Name N'-(2-aminoethyl)ethane-1,2-diamine
Viscosity 5.05 mPa.s (20°C)
Formula Weight 103.17
Percent Purity 98+%
Physical Form Liquid
Chemical Name or Material Diethylenetriamine
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Harmful in contact with skin.
Fatal if inhaled.
Harmful if swallowed.
May cause an allergic skin reaction.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF SWALLOWED: rinse mouth
EINECSNumber 203-865-4

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.