Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Amino-2-nitrophenol, 99%

Catalog No. AC164230050
Change view
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC164230050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC164230050 Supplier Thermo Scientific Chemicals Supplier No. 164230050
Add to Cart

CAS: 119-34-6 | C6H6N2O3 | 154.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 119-34-6
Molecular Formula C6H6N2O3
Molecular Weight (g/mol) 154.12
MDL Number MFCD00007876
InChI Key WHODQVWERNSQEO-UHFFFAOYSA-N
Synonym phenol, 4-amino-2-nitro, fourrine 57, 4-hydroxy-3-nitroaniline, 2-nitro-4-aminophenol, fourrine brown pr, oxidation base 25, fourrine brown propyl, o-nitro-p-aminophenol, p-amino-o-nitrophenol, c.i. oxidation base 25
PubChem CID 3417419
ChEBI CHEBI:82385
IUPAC Name 4-amino-2-nitrophenol
SMILES C1=CC(=C(C=C1N)[N+](=O)[O-])O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 124.0°C to 128.0°C
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula H2NC6H3(NO2)OH
Quantity 5 g
Beilstein 13, 520
Solubility Information Solubility in water: .. Other solubilities: soluble in diethylether, ethanol and aceton
Formula Weight 154.12
Percent Purity 99%
Chemical Name or Material 4-Amino-2-nitrophenol, 99%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes serious eye damage.
Suspected of causing genetic defects.
Causes skin irritation.
May cause respiratory irritation.
Harmful if swallowed.
Harmful in contact with skin.
Precautionary Statement GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
Take off contaminated clothing and wash before reuse.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Con
EINECSNumber 204-316-1

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.