Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Seamless Cellulose Dialysis Tubing
SCI_ED

Catalog No. S25645F Shop All Fisherbrand Products
Change view
Click to view available options
Diameter (Metric):
6.4 mm
15.9 mm
20.4 mm
28.6 mm
32 mm
45 mm
47.7 mm
Length (Metric):
3 m
30 m
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Diameter (Metric) Length (Metric)
S25645F 15.9 mm 30 m
S25645B 28.6 mm 3 m
S25645A 20.4 mm 3 m
S25645 15.9 mm 3 m
S25645C 47.7 mm 3 m
S25645D 6.4 mm 3 m
S25645E 6.4 mm 30 m
S25645H 45 mm 30 m
S25645G 32 mm 30 m
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Catalog No. S25645F
Only null left
Add to Cart
Add to Cart

Reliable and flexible dialysis tubing with 12,000 MWCO for separating macromolecules in educational chemistry, biology, and pharmaceutical labs.

  • Material: Seamless cellulose
  • Molecular Weight Cut-Off (MWCO): Retains molecules ≥12,000
  • Da Applications: Ideal for educational settings in analytical chemistry, biology, and pharmaceutical studies
  • Permeability: High permeability to water and low molecular weight compounds
  • Pore Diameter: Approx. 4.8 nm
  • Formulation: Glycerin, water, and 1% sulfur
  • Storage: Store in an airtight container; rehumidify if dried out
  • Diameter: 45 mm (1.13 in.)
  • Length: 30 m (100 ft.)
  • Quantity: Sold individually (Each)
  • Educational Use Only: No Certificate of Analysis (C of A) provided

This product is for educational use - no C of A is available.

Specifications

Product Type Dialysis Tubing
Material Seamless Cellulose
Diameter (Metric) 15.9 mm
Diameter (English) 0.62 in.
Length (English) 100 ft.
Length (Metric) 30 m
Formulation Glycerin, water and 1% sulfur
Quantity Each
For Use With (Application) Useful in analytical chemistry, biology, and pharmaceutical fields
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.