Learn More
Diagenode Inc TUBES TPX 1.5ML 50/PK
Supplier:  Diagenode Inc C3001001050
Encompass
Tubes Diagenode 1.5ml, pack of 50 TPX Polymethylpentene (PMP) tubes improve sonication and shearing efficiency. TPX is a hard plastic which improves the transmission of the ultrasound to the sample. TPX tubes have been validated for use on the Bioruptor Standard and Bioruptor Plus. Recommended sample volume: 100 ul - 300 ul.
Catalog No. NC0065146