Learn More
Tecan 200L MCA96 Tip NFil
Supplier:  Tecan 30038617
Encompass_Preferred
Genuine Tecan 200 L disposable automation tips designed specifically for use on the Freedom EVO and Fluent MultiChannel Arm™ (MCA) 96 option. ANSI format, single stack, non-filtered. Tecan STERILE purity level - sterile, tested and certified to be free from human DNA, Dnase, Rnase, pyrogens and endotoxins. 40 tip boxes x 96 tips, 3,840 tips/sales unit. All MultiChannel Arm™(MCA) 96 disposable tips are designed, manufactured, tested and certified to guarantee reliable quality, helping you to acquire optimal results.
Catalog No. NC0080816
Add to Cart
missing translation for 'nonReturnableMessage' missing translation for 'returnPolicyLabel'