Learn More
Lonza Walkersville MycoAlert PLUS Mycoplasma Detection Kit (10 Tests)
Catalog No. NC0395309
Encompass
Lonza’s MycoAlert Mycoplasma Detection Assays are selective biochemical tests to detect Mycoplasma contamination in cell cultures. They exploit the activity of mycoplasmal enzymes which are found in the vast majority of about 200 mycoplasma species, but are not present in eukaryotic cells.Benefits:Easy assay: Simply add two reagents to the culture supernatant and perform two luminescence readingsResults in < 20 minEasy interpretation of resultsApplications:Detects all common mollicute contaminations (Mycoplasma, Acholeplasma, Entomoplasma and Spiroplasma; except Ureaplasma)Suited for tube luminometers, plate luminometers and multifunctional readersContent:1 x 1.2 mL MycoAlert PLUS Reagent (lyophilized)1 x 1.2 mL MycoAlert PLUS Substrate (lyophilized)1 x 3 mL MycoAlert PLUS Assay Buffer
Catalog No. NC0395309 Supplier Lonza Walkersville Supplier No. LT07701
Lonza’s MycoAlert Mycoplasma Detection Assays are selective biochemical tests to detect Mycoplasma contamination in cell cultures. They exploit the activity of mycoplasmal enzymes which are found in the vast majority of about 200 mycoplasma species, but are not present in eukaryotic cells.Benefits:Easy assay: Simply add two reagents to the culture supernatant and perform two luminescence readingsResults in < 20 minEasy interpretation of resultsApplications:Detects all common mollicute contaminations (Mycoplasma, Acholeplasma, Entomoplasma and Spiroplasma; except Ureaplasma)Suited for tube luminometers, plate luminometers and multifunctional readersContent:1 x 1.2 mL MycoAlert PLUS Reagent (lyophilized)1 x 1.2 mL MycoAlert PLUS Substrate (lyophilized)1 x 3 mL MycoAlert PLUS Assay Buffer