Learn More
New England Biolabs, Inc. PNGase A – 150 units
SDP
Supplier:  New England Biolabs, Inc. P0707S
Encompass
PNGase A cleaves between the innermost GlcNAc and asparagine residues of high mannose, hybrid, and short complex oligosaccharides such as those found in plant and insect cells from N-linked glycoproteins and glycopeptides. PNGase A differs from PNGase F in that it cleaves N-linked glycans with or without (1,3)-linked core fucose residues.
Catalog No. NC1193434