Learn More
101 BIO Lentivirus 10X Titer-Up Reagent | 1 ml, for packaging 500 ml lentivirus soup
Catalog No. nc1792390
Encompass
This virus titer boost is a novel recipe of small molecules designed for increasing / boosting lentivirus packaging efficiency. Lentivirus 10X Titer-Up stably regulates the viral RNA packaging on the transcriptional level, which can enhance production of lentiviral particles for up to 10 fold.
Catalog No. NC1792390 Supplier 101 BIO Supplier No. P906
This virus titer boost is a novel recipe of small molecules designed for increasing / boosting lentivirus packaging efficiency. Lentivirus 10X Titer-Up stably regulates the viral RNA packaging on the transcriptional level, which can enhance production of lentiviral particles for up to 10 fold.