Learn More
Lascar Electronics Inc High Accuracy Temp & Relative Humidity Data Logger with USB
SDP
Supplier:  Lascar Electronics Inc ELUSB2+
Encompass_Preferred
This standalone data logger measures over 16,000 readings over a -35 to +80°C (-31 to +176°­­F) and 0 to 100% humidity (RH) range. Easily set up the logger and view downloaded data by plugging the unit into a PC’s USB port and using the free EasyLog software provided. Data can then be graphed, printed and exported to other applications for detailed analysis. Supplied with ½ AA battery and wall mount clip.
Catalog No. NC1856443