Learn More
Chemglass Life Sciences Tripod Stand for 1-3 L Jacketed Bioreactor Vessels
SDP
Supplier:  Chemglass Life Sciences CLS1381STD3
Encompass
Tripod stand designed to support jacketed bioreactor vessels in the 1-3 L range. The top two-piece ring is anodized aluminum, while the lower ring and legs are electropolished stainless steel. The stand is supplied complete with a silicone o-ring and knurled nuts; optional stabilizer bars are available for added stability.
  • Supports jacketed bioreactor vessels in the 1-3 L capacity range.
  • Top two-piece ring constructed from anodized aluminum.
  • Lower ring and legs made from electropolished stainless steel.
  • Supplied with silicone o-ring and six knurled nuts for head plate attachment.
  • Optional stabilizer bars attach to legs for extra stability.
Catalog No. NC1864950