Learn More
Vector Laboratories Lycopersicon Esculentum (Tomato) Lectin (LEL, TL), DyLight 649 | 1mg
SDP
Supplier:  Vector Laboratories DL11781
Encompass
Tomato lectin (from Lycopersicon esculentum) is an effective marker of blood vessels and microglial cells in rodents. Conjugation of the lectin with a fluorophore facilitates fast, one-step detection and visualization using intravascular perfusion methods or direct application to tissue sections. DyLight 649 labeled tomato lectin has an appropriate number of fluorochromes bound to provide the optimum staining characteristics for this lectin. This conjugate is supplied essentially free of unconjugated fluorochromes.
Catalog No. NC1866411