Learn More
Teknova 10% Tween-20. 200mL, Dnase/Rnase Tested, Sterile.
SDP
Supplier:  Teknova T0027
Encompass
Tween Solution is a nonionic detergent used for protein extraction and isolation of nuclei from mammalian cell lines. It is soluble in water and is widely used in biochemical applications such as solubilizing proteins, isolating nuclei from cells, and emulsifying and dispersing substances in medicinal and food products.
Catalog No. NC2401211