Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PFEIFFER VACUUM INC Pfeiffer Vacuum Mineral Oil, P3, Hydrocarbon, 0.5 L, Oxidation-Resistant

Supplier:  PFEIFFER VACUUM INC 2000025868

Encompass_Preferred

Pfeiffer Vacuum P3 Mineral Oil is a high-quality hydrocarbon oil specifically designed for use with rotary vane vacuum pumps in standard applications, including pumping air, inert gases, and noble gases. It is formulated to be oxidation-resistant and suitable for high operating temperatures up to a maximum of 95°C.

  • Designed for standard applications
  • Suitable for pumping air, inert gases, and noble gases
  • Oxidation-resistant
  • Withstands operating temperatures up to 95°C
  • Achieves a final pressure of < 1 · 10-3 hPa
  • Available in 0.5 L volume

Catalog No. NC3979035


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.