Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ACROBiosystems ACRO BIOSYSTEMS Glycoprotein B, Biotinylated, HSV-2 (strain 333), His,Avitag

Catalog No. NC3989512
Encompass_Preferred

Envelope glycoprotein that forms spikes at the surface of the virion envelope. It is essential for the initial attachment to heparan sulfate moieties of host cell surface proteoglycans. Involved in the fusion of viral and cellular membranes, leading to virus entry into the host cell. Membrane fusion is mediated by the fusion machinery, including gB and the heterodimer gH/gL, following initial binding to host receptors.

  • Expressed from human 293 cells (HEK293).
  • Features a polyhistidine tag and an Avi tag (Avitag™) at the C-terminus.
  • Biotinylated using Avitag™ technology.
  • Designed as a trimer with a calculated molecular weight of 76.0 kDa.
  • Purity greater than 90% as determined by SDS-PAGE and SEC-MALS.
  • Bioactivity confirmed by ELISA, binding mouse anti-herpes virus 1 and 2.
  • Store at -70°C or lower; avoid repeated freeze-thaw cycles.

Catalog No. NC3989512 Supplier ACROBiosystems Supplier No. GLBV82E3100UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Envelope glycoprotein that forms spikes at the surface of the virion envelope. It is essential for the initial attachment to heparan sulfate moieties of host cell surface proteoglycans. Involved in the fusion of viral and cellular membranes, leading to virus entry into the host cell. Membrane fusion is mediated by the fusion machinery, including gB and the heterodimer gH/gL, following initial binding to host receptors.

  • Expressed from human 293 cells (HEK293).
  • Features a polyhistidine tag and an Avi tag (Avitag™) at the C-terminus.
  • Biotinylated using Avitag™ technology.
  • Designed as a trimer with a calculated molecular weight of 76.0 kDa.
  • Purity greater than 90% as determined by SDS-PAGE and SEC-MALS.
  • Bioactivity confirmed by ELISA, binding mouse anti-herpes virus 1 and 2.
  • Store at -70°C or lower; avoid repeated freeze-thaw cycles.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.