Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Edvotek™ A-maize-ing Editing: Using CRISPR to Improve Crops Kit
SDPSCI_ED

Catalog No. s27553 Shop All EDVOTEK Products
Change view
Click to view available options
Class Size:
8 Lab Groups
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S27553 8 Lab Groups
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S27553 Supplier Edvotek™ Supplier No. 210
Only null left
Add to Cart
Add to Cart

Students will assume the role of plant geneticists working to develop corn crops capable of thriving in a changing environment. This experiment will allow students to develop an understanding of CRISPR-Cas9 applications, cleave DNA, and examine results after gel electrophoresis.

  • Some Components Require Freezer Storage Upon Receipt
  • Sufficent for 8 Gels
  • Time Range: 80 min. to 105 min.

Specifications

Grade 9 to Undergraduate
Age 14 to 20
Includes Instructions, Maize DNA samples, DNA Standard Marker, Simulated sgRNAs and Cas9 enzyme, CRISPR Reaction Buffer, Reconstitution Buffer, UltraSpec-Agarose™, 10 x Gel Loading Solution, 50 x Concentrated Electrophoresis Buffer, Practice Gel Loading Solution, FlashBlue™ DNA Stain, and Microcentrifuge Tubes
Requires DNA Electrophoresis, Micropipettes: 5-50 μL (Optional), Microwave/Hot Plate
Time Range 80 min. to 105 min.
Content And Storage Some Components Require Freezer Storage Upon Receipt
Class Size 8 Lab Groups
Product Type Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.