Learn More
MilliporeSigma™ E-POD™ Remote Dispenser
Catalog No. ziqep0d00 Shop All MilliporeSigma Products
Change view
Click to view available options
:
E-POD™ Remote Dispenser
Catalog No.
ZIQEP0D00 E-POD™ Remote Dispenser
1 options
Catalog No. ZIQEP0D00 Supplier MilliporeSigma™ Supplier No. ZIQEP0D00
Only null left
Add to Cart
Add to Cart
The most integrated pure and ultrapure water system that is intelligent, intuitive and green.
  • Intelligent pure water dispenser.
  • 1 E-POD™s can be connected to a Milli-Q™ IQ 7003/5 water purification system.
TRUSTED_SUSTAINABILITY
Specifications
Capacity ≤30 ppb
For Use With (Equipment) Intelligent pure water dispenser. 1 E-POD™ can be connected to a Milli-Q™ IQ 7003/5 water purification system.
Includes E-POD™ remote dispenser
Type E-POD™ Remote Dispenser
Length (Metric) 70.2 cm
Width (Metric) 21 cm
Height (Metric) 70.2 cm