Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bismuth(III) oxide, 99%

Catalog No. AA12230A7
Change view
Click to view available options
Quantity:
250 g
1 kg
5 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA12230A7 5 kg
AA1223030 250 g
AA12230A1 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA12230A7 missing translation for 'mfr' Thermo Scientific Chemicals missing translation for 'supplierNo' 012230A7
Only null left
Add to Cart

CAS: 1304-76-3 | Bi2O3 | 465.96 g/mol

It finds use in disinfectants, magnets, glass, rubber, vulcanization, fireproofing papers and polymers, and catalysts. Bismuth trioxide brings about the dragon's eggs effect in fireworks, as a replacement of red lead. Bismuth(III) compounds are attractive reagents and catalysts in organic synthesis because of their low cost and ease of handling. Bismuth trioxide nanoparticles also play an important role in high energy gas generator. The alpha crystalline form of bismuth(III) oxide has p-type electronic conductivity.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1304-76-3
Molecular Formula Bi2O3
Molecular Weight (g/mol) 465.96
MDL Number MFCD00003462
InChI Key TYIXMATWDRGMPF-UHFFFAOYSA-N
IUPAC Name dibismuth(3+) trioxidandiide
SMILES [O--].[O--].[O--].[Bi+3].[Bi+3]

Specifications

Boiling Point ca 1890°C
Melting Point 817°C
Color White
Physical Form Powder
Quantity 5 kg
Assay Percent Range 99%
Solubility Information Practically insoluble in water. Soluble in acids.
Formula Weight 465.96
Density 8.9
Chemical Name or Material Bismuth(III)
missing translation for 'healthHazard1' H303: May be harmful if swallowed.
EINECSNumber 215-134-7
RTECSNumber EB2984460
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.