Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

9-Borabicyclo[3.3.1]nonane, 0.5M solution in THF, AcroSeal™

Catalog No. AC165811000
Click to view available options
Quantity:
100 mL
800 mL
Packaging:
AcroSeal™ Glass bottle

Catalog No. AC165811000

Supplier: Thermo Scientific Chemicals 165811000

Only null left
Add to Cart

CAS: 280-64-8 | C8H15B | 122.02 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Lewis acid, Reduction reagent
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 9-Borabicyclo[3.3.1]nonane
CAS 280-64-8, 109-99-9
CAS Min % 6.14, 92.49
CAS Max % 7.51, 93.86
Density 0.8840g/mL
Flash Point −17°C
Packaging AcroSeal™ Glass bottle
Molecular Formula C8H15B
Quantity 100 mL
MDL Number MFCD00074742
Fieser 02,31; 03,24; 04,41; 05,46; 06,62; 07,29; 08,47; 09,57; 10,48; 11,68; 13,92; 14,52; 15,43; 17,49; 01,732; 04,356
Specific Gravity 0.884
Synonym 9-borabicyclo 3.3.1 nonane, 9-borabicyclo 3.3.1 nonan, 9-borabicyclo 3,3,1 nonane, 9$l^ 2-borabicyclo 3.3.1 nonane, 1s,5s-9-borabicyclo 3.3.1 nonane, monomer; 21205-91-4 dimer, 9-borabicyclo 3.3.1 nonane,9-bbn,intetrahydrofuran
Solubility Information Solubility in water: reacts
InChI Key AMKGKYQBASDDJB-UHFFFAOYSA-N
SMILES [B]1C2CCCC1CCC2
IUPAC Name 9$l^{2}-borabicyclo[3.3.1]nonane
Molecular Weight (g/mol) 122.02
PubChem CID 6327450
Formula Weight 122.02
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.